ContactUsLinkComponent
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Certificates
undefined
Potassium xanthogenate (potassium-O-ethyl dithiocarbonate) 97+%
Specification Test Results
  • Appearance:
    Yellow Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 %
  • Titration with HClO4:
    ≥97.0 % (On dry substance)