FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) ≥90% (by HPLC), powder suitable for protein labeling, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:3326-32-7
:
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063
:389.381 g/mol
Melting Pt:>360 °C (lit.)
:M-Clarity™ quality level = MQ200
EINECS:222-042-0
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Appearance (Colour):Orange to Dark Orange
- Appearance (Form):Powder
- Solubility (Colour):Light Green-Yellow to Very Dark Green-Yellow and Faint Yellow to Dark Yellow
- Solubility (Turbidity), 1 mg/ml, Acetone:Clear
- Proton NMR Spectrum:Conforms to Structure
- Degree of Coupling:≥85.0%
- Purity (HPLC):≥90%
- Recommended Retest Period:2 years
Specification Test Results
Frequently Bought Together