Order Entry
Austria
ContactUsLinkComponent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Irritant
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # PIER46425
CAS Number:  
Irritant
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)
Catalog # PIER46425
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :389.381 g/mol
EINECS:222-042-0
 :GHS07
Storage Temperature:Refrigerator
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063