
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I), Sigma-Aldrich®
Catalog # 1.24546.0000
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Solubility:(20 °C) insoluble
- Melting Point:360 °C
- Molar Mass:389.38 g/mol
Specification Test Results
About this item
:
:389.381 g/mol
EINECS:222-042-0
:GHS07
Storage Temperature:Refrigerator
:1
CAS Number:3326-32-7
Formula:C₂₁H₁₁NO₅S
MDL Number:MFCD00005063