Deoxy-big CHAP (N,N-bis[3-(D-gluconamido)propyl]deoxycholamide)
Catalog # DG026
Supplier: G-Biosciences
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:862.05 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:86303-23-3
Formula:C₄₂H₇₅N₃O₁₅
MDL Number:MFCD00161482