Order Entry
Austria
ContactUsLinkComponent
Azure B (azure I) ≥85% (by HPLC), prepared by direct synthesis, Sigma-Aldrich®
Irritant
Azure B (azure I) ≥85% (by HPLC), prepared by direct synthesis, Sigma-Aldrich®
Certificates
undefined
Irritant
Azure B (azure I) ≥85% (by HPLC), prepared by direct synthesis, Sigma-Aldrich®

Specification Test Results

  • Appearance (Form):
    Powder
  • Infrared spectrum:
    Conforms to Structure
  • Absorbance:
    638 - 644
  • Extinction Coefficient:
    ≥67000
  • Carbon:
    ≥49.0 %
  • Nitrogen:
    ≥11.4 %
  • Purity (HPLC):
    ≥85.0 %
  • Solubility (Color), c = 1mg/ml; Water:
    Blue to Very Dark Blue

Specification Test Results

Frequently Bought Together