Order Entry
Austria
ContactUsLinkComponent
 
Streptavidin
  ROCKS000-01
 :  
undefined
Streptavidin
  ROCKS000-01
 :  

 

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 mg
  • CAS No.:
    9013-20-1
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00082035
  • Protein/peptide name:
    Streptavidin