Streptavidin
Plastic bottle for solids, 1 mg
Catalog # NEWEN7021S
Marketsource
Supplier: NEW ENGLAND BIOLABS
Jump to:
Product Details & Documents
Specifications
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 mg
- Cat. No.:NEWEN7021S
- Protein/peptide name:Streptavidin
- Storage conditions:Freezer
- MDL No.:MFCD00082035
- CAS No.:9013-20-1



