Order Entry
Austria
ContactUsLinkComponent
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), Millipore®
Corrosive
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), Millipore®
Catalog # 580560-1
CAS Number:  
Corrosive
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride), Millipore®
Catalog # 580560-1
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :286.65 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
 :GHS05
CAS Number:51805-45-9
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469