Bovine aprotinin
786-046
:
- Pk:100 mg
- CAS No.:9087-70-1
- Species:Bovine
- Storage conditions:Refrigerator
- MDL No.:MFCD00130541
- Protein synonyms:Trypsin inhibitor, pancreatic basic
- Protein/peptide name:Aprotinin
- Molecular formula:C₂₈₄H₄₃₂N₈₄O₇₉S₇
Aprotinin is a broad range, competitive and reversible inhibitor of chymotrypsin, plasmin, trypsin, kallikrein and other serine proteases. Activity: approx 6,000 KIU/mg.