Aprotinin
Supplier: PanReac AppliChem
Certificates
About this item
Specifications
- CAS No.:9087-70-1
- Storage conditions:Refrigerator
- MDL No.:MFCD00130541
- Protein synonyms:Trypsin inhibitor, pancreatic basic
- Protein/peptide name:Aprotinin
- Molecular formula:C₂₈₄H₄₃₂N₈₄O₇₉S₇
Specifications
Frequently Bought Together