Oil red O, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:1320-06-5
:
Formula:C₂₆H₂₄N₄O
MDL Number:MFCD00003898
:408.51 g/mol
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:215-295-3
Related Information
Frequently Bought Together