Lithium methoxide 1 mol/l in methanol, Sigma-Aldrich®
: Merck
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Nitrile Gloves
: VWR Collection
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Accessories for Mettler balances
: Mettler - Toledo
Ethanol 96% ≥96% (v/v), TechniSolv®, pure
: VWR Chemicals
Disinfectant solutions, Bacillol AF
: BODE CHEMIE
Formaldehyde 36% in aqueous solution stabilised, GPR RECTAPUR®
: VWR Chemicals