Order Entry
Austria
ContactUsLinkComponent
Hydroxyethyl cellulose for synthesis, Sigma-Aldrich®
Hydroxyethyl cellulose for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Hydroxyethyl cellulose for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates

Specification Test Results

  • Assay (cellulose ether):
    ≥92%
  • Bulk Density:
    350 kg/m3
  • pH:
    (10 g/l, H₂O, 20 °C) neutral
  • Ignition Point:
    >360 °C

Specification Test Results

Related Information

Frequently Bought Together