Order Entry
Austria
ContactUsLinkComponent
Hydroxyethyl cellulose, high purity
Hydroxyethyl cellulose, high purity
Catalog # K391-500G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydroxyethyl cellulose, high purity
Catalog # K391-500G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Bottle, amber
  • Pk:
    500 g
  • Seq:
    3645

Specifications

Specification Test Results

  • pH (2%, water) @25 °C:
    6 – 7

Specification Test Results

About this item

 :
 :GHS07
Storage Temperature:Ambient
CAS Number:9004-62-0
MDL Number:MFCD00072770