X-GlcA ((5-bromo-4-chloro-3-indolyl-ß-D-glucuronide) cyclohexylammonium salt), Sigma-Aldrich®
Catalog # 203783-100
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:521.79 g/mol
Melting Pt:230 °C
Storage Temperature:Freezer
CAS Number:114162-64-0
Formula:C₂₀H₂₆BrClN₂O₇
MDL Number:MFCD00058543