3,3',5,5'-Tetramethylbenzidine (TMB), insoluble, Calbiochem®, Sigma-Aldrich®
Catalog # 613548-100
Supplier: Merck Millipore (Oncogene)
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 mL
- Seq:2440
- Environmentally Preferable:
Specifications
About this item
Synonyms: TMB, 3,3',5,5'-Tetramethyl-[1,1'-biphenyl]-4,4'-diamine
:
:240.35 g/mol
Boiling Pt:402
Melting Pt:166…170 °C
Storage Temperature:Ambient
EINECS:259-364-6
CAS Number:54827-17-7
Formula:C₁₆H₂₀N₂
MDL Number:MFCD00007748