Order Entry
Austria
ContactUsLinkComponent
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn) standard for AAS and ICP
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn) standard for AAS and ICP
Catalog # PERCN9300161
Supplier:  PerkinElmer
CAS Number:  
Tin standard solution, 1000 mg/L Sn in 20% hydrochloric acid (from Sn) standard for AAS and ICP
Catalog # PERCN9300161
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    20% HCl
  • Pk:
    1
  • Element:
    Tin
  • Symbol:
    Sn

Specifications

About this item