Order Entry
Austria
ContactUsLinkComponent
 
Potassium standard solution, 1,000 mg/L K in dilute nitric acid (from KNO₃) standard for AAS and ICP
  PERCN9300141
 :  
Potassium standard solution, 1,000 mg/L K in dilute nitric acid (from KNO₃) standard for AAS and ICP
  PERCN9300141
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Pk Info:
    1× 500 ml
  • Element:
    Potassium
  • Symbol:
    K