Order Entry
Austria
ContactUsLinkComponent
 
Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid (de Ho₂O₃) standard for AAS and ICP
  PERCN9300123
 :  
Holmium standard solution, 1,000 mg/L Ho in dilute nitric acid (de Ho₂O₃) standard for AAS and ICP
  PERCN9300123
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Holmium
  • Symbol:
    Ho