Calcium standard solution, 1,000 mg/L Ca in dil. nitric acid (from CaCO3) standard for AAS and ICP
Catalog # PERCN9300108
Supplier: PerkinElmer
CAS Number:
Specifications
- Concentration:1000 mg/L
- Matrix:2% HNO₃
- Pk:1
- Element:Calcium
- Symbol:Ca