Order Entry
Austria
ContactUsLinkComponent
 
Terbium standard solution, 1000 mg/L Tb in dil. nitric acid (from Tb4O7) standard for AAS and ICP
Catalog # PERCN9300157
Supplier:  PerkinElmer
CAS Number:  
Terbium standard solution, 1000 mg/L Tb in dil. nitric acid (from Tb4O7) standard for AAS and ICP
Catalog # PERCN9300157
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Terbium
  • Symbol:
    Tb

Specifications

About this item