Order Entry
Austria
ContactUsLinkComponent
Rhodium standard solution, 1,000 mg/L Rh in 10% hydrochloric acid (from RhCl3) standard for AAS and ICP
Rhodium standard solution, 1,000 mg/L Rh in 10% hydrochloric acid (from RhCl3) standard for AAS and ICP
Catalog # PERCN9300144
Supplier:  PerkinElmer
CAS Number:  
Rhodium standard solution, 1,000 mg/L Rh in 10% hydrochloric acid (from RhCl3) standard for AAS and ICP
Catalog # PERCN9300144
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pk:
    1
  • Pk Info:
    1× 500 ml
  • Element:
    Rhodium
  • Symbol:
    Rh

Specifications

About this item