Order Entry
Austria
ContactUsLinkComponent
Palladium standard solution, 1,000 mg/L Pd in 10% hydrochloric acid (from (NH4)2PdCl4) standard for AAS and ICP
Palladium standard solution, 1,000 mg/L Pd in 10% hydrochloric acid (from (NH4)2PdCl4) standard for AAS and ICP
Catalog # PERCN9300138
Supplier:  PerkinElmer
CAS Number:  
Palladium standard solution, 1,000 mg/L Pd in 10% hydrochloric acid (from (NH4)2PdCl4) standard for AAS and ICP
Catalog # PERCN9300138
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pk:
    1
  • Pk Info:
    1× 500 ml
  • Element:
    Palladium
  • Symbol:
    Pd

Specifications

About this item