Order Entry
Austria
ContactUsLinkComponent
Sodium standard solution, 1000 mg/L Na in dil. nitric acid (from Na₂CO₃) standard for AAS and ICP
Sodium standard solution, 1000 mg/L Na in dil. nitric acid (from Na₂CO₃) standard for AAS and ICP
Catalog # PERCN9300152
Supplier:  PerkinElmer
CAS Number:  
Sodium standard solution, 1000 mg/L Na in dil. nitric acid (from Na₂CO₃) standard for AAS and ICP
Catalog # PERCN9300152
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Sodium
  • Symbol:
    Na

Specifications

About this item