Order Entry
Austria
ContactUsLinkComponent
 
Cobalt standard solution, 1,000 mg/L Co in dil. nitric acid (from CoCO3) standard for AAS and ICP
Catalog # PERCN9300113
Supplier:  PerkinElmer
CAS Number:  
Cobalt standard solution, 1,000 mg/L Co in dil. nitric acid (from CoCO3) standard for AAS and ICP
Catalog # PERCN9300113
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Cobalt
  • Symbol:
    Co

Specifications

About this item