Order Entry
Austria
ContactUsLinkComponent
 
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄) standard for AAS and ICP
  PERCN9300154
 :  
Sulphur standard solution, 1000 mg/L S in water (from (NH₄)₂SO₄) standard for AAS and ICP
  PERCN9300154
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    H₂O
  • Pk:
    1
  • Symbol:
    S