Order Entry
Austria
ContactUsLinkComponent
Cesium standard solution, 1,000 mg/L Cs in dil. nitric acid (from CsNO₃) standard for AAS and ICP
Cesium standard solution, 1,000 mg/L Cs in dil. nitric acid (from CsNO₃) standard for AAS and ICP
Catalog # PERCN9300111
Supplier:  PerkinElmer
CAS Number:  
Cesium standard solution, 1,000 mg/L Cs in dil. nitric acid (from CsNO₃) standard for AAS and ICP
Catalog # PERCN9300111
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Cesium
  • Symbol:
    Cs

Specifications

About this item