Order Entry
Austria
ContactUsLinkComponent
Manganese standard solution, 1000 mg/L Mn in dil. nitric acid (from Mn) standard for AAS and ICP
Manganese standard solution, 1000 mg/L Mn in dil. nitric acid (from Mn) standard for AAS and ICP
Catalog # PERCN9300132
Supplier:  PerkinElmer
CAS Number:  
Manganese standard solution, 1000 mg/L Mn in dil. nitric acid (from Mn) standard for AAS and ICP
Catalog # PERCN9300132
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    500 mL
  • Element:
    Manganese
  • Symbol:
    Mn

Specifications

About this item