ContactUsLinkComponent
Microfiltration filters, MiniKap®
Microfiltration filters, MiniKap®
Catalog # SPECMK2M204V6N
Microfiltration filters, MiniKap®
Catalog # SPECMK2M204V6N
Specifications
  • Filtration area:
    225 cm²
  • Pore size:
    0,2 µm
  • Type:
    Microfiltration filters, MiniKap®
  • Inlet/outlet:
    MNPT 1/4 inches / MNPT 1/4 inches
  • Sample volume:
    500 - 1000 ml
  • Pk:
    6
Specifications
About this item
These Point-of-use microfiltration filters are excellent for water and gas filtration systems. MiniKap® HF filter provides the filtration efficiency of the 0,2 µm DynaFibre® and air vent fibre in a device conveniently equipped with a ¼'' MNPT, Luer-Lok or hose barb inlet connection.