Orders LinkContactUsLinkComponent
Liquid sampling dippers
Liquid sampling dippers
Supplier:   Sampling Systems
Liquid sampling dippers
Supplier:  Sampling Systems
Certificates
Liquid sampling dippers
Liquid sampling dippers
9 Options Available Below
Product Details & Documents
Stainless dippers are ideal for taking surface samples from liquids and suspensions. The sampler has a 316 L stainless steel cup welded to a long reach handle. The handles are made from strong solid stainless steel bar.
  • Range of cup sizes from 100 to 500 ml
  • Lengths from 600 to 1500 mm
  • 316 L stainless steel
  • Polished finish
To take a sample you simply insert the dipper into the liquid to take a surface sample. They are ideal for obtaining samples from a wide range of environments including use in pharmaceutical, chemical and food industries.
Product Family Options
Product Information
  • Material
    Length
    Capacity
    Availability
    Price
  • SAMP5005A-100
    Sample dipper
    Sample dipper
    Stainless steel 316
    600 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 157,00 €
    157,00 €
    1 * 1 ST
       157,00 €
  • SAMP5005A-250
    Sample dipper
    Sample dipper
    Stainless steel 316
    600 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 161,00 €
    161,00 €
    1 * 1 ST
       161,00 €
  • SAMP5005A-500
    Sample dipper
    Sample dipper
    Stainless steel 316
    600 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 186,00 €
    186,00 €
    1 * 1 ST
       186,00 €
  • SAMP5005B-100
    Sample dipper
    Sample dipper
    Stainless steel 316
    1000 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 189,00 €
    189,00 €
    1 * 1 ST
       189,00 €
  • SAMP5005B-250
    Sample dipper
    Sample dipper
    Stainless steel 316
    1000 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 218,00 €
    218,00 €
    1 * 1 ST
       218,00 €
  • SAMP5005B-500
    Sample dipper
    Sample dipper
    Stainless steel 316
    1000 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 269,00 €
    269,00 €
    1 * 1 ST
       269,00 €
  • 300-0461
    Sample dipper
    Sample dipper
    Stainless steel 316
    1500 mm
    100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 257,00 €
    257,00 €
    1 * 1 ST
       257,00 €
  • 300-0462
    Sample dipper
    Sample dipper
    Stainless steel 316
    1500 mm
    250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 252,00 €
    252,00 €
    1 * 1 ST
       252,00 €
  • 300-0463
    Sample dipper
    Sample dipper
    Stainless steel 316
    1500 mm
    500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 281,00 €
    281,00 €
    1 * 1 ST
       281,00 €