Orders LinkContactUsLinkComponent
Samplers, U-D Master
Supplier:   Sampling Systems
Samplers, U-D Master
Supplier:  Sampling Systems
Certificates
Samplers, U-D Master
15 Options Available Below
Product Details & Documents
U-D Master is designed for sampling free flowing powders. By simply changing the sample insert cells, different volumes can be sampled.
  • Operation of the unit dose powder sampler is very easy
  • Push the sampler: At the required depth rotate the handle, which will take the sample
  • After removing the sampler the sample inserts can be removed for easy emptying of the sample
Single version takes one sample from each location.
Double version takes two samples from each location.
Triple version takes three samples from each location.
Product Family Options
Product Information
  • Length
    No. of chambers
    Ø
    Availability
    Price
  • SAMP1230A-300
    Single style sampler
    Single style sampler
    300 mm
    5
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 530,00 €
    4 530,00 €
    1 * 1 ST
       4 530,00 €
  • SAMP1230A-600
    Single style sampler
    Single style sampler
    600 mm
    10
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 400,00 €
    5 400,00 €
    1 * 1 ST
       5 400,00 €
  • SAMP1230A-900
    Single style sampler
    Single style sampler
    900 mm
    6
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 810,00 €
    5 810,00 €
    1 * 1 ST
       5 810,00 €
  • SAMP1230A-1200
    Single style sampler
    Single style sampler
    1200 mm
    8
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 350,00 €
    6 350,00 €
    1 * 1 ST
       6 350,00 €
  • SAMP1230A-1500
    Single style sampler
    Single style sampler
    1500 mm
    10
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 7 180,00 €
    7 180,00 €
    1 * 1 ST
       7 180,00 €
  • SAMP1230A-1800
    Single style sampler
    Single style sampler
    1800 mm
    12
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 240,00 €
    8 240,00 €
    1 * 1 ST
       8 240,00 €
  • SAMP1230B-600
    Double style sampler
    Double style sampler
    600 mm
    5
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 6 350,00 €
    6 350,00 €
    1 * 1 ST
       6 350,00 €
  • SAMP1230B-900
    Double style sampler
    Double style sampler
    900 mm
    7
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 8 240,00 €
    8 240,00 €
    1 * 1 ST
       8 240,00 €
  • SAMP1230B-1200
    Double style sampler
    Double style sampler
    1200 mm
    8
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9 190,00 €
    9 190,00 €
    1 * 1 ST
       9 190,00 €
  • SAMP1230B-1500
    Double style sampler
    Double style sampler
    1500 mm
    10
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10 040,00 €
    10 040,00 €
    1 * 1 ST
       10 040,00 €
  • SAMP1230B-1800
    Double style sampler
    Double style sampler
    1800 mm
    12
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10 970,00 €
    10 970,00 €
    1 * 1 ST
       10 970,00 €
  • SAMP1230C-900
    Triple style sampler
    Triple style sampler
    900 mm
    5
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9 570,00 €
    9 570,00 €
    1 * 1 ST
       9 570,00 €
  • SAMP1230C-1200
    Triple style sampler
    Triple style sampler
    1200 mm
    7
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 10 970,00 €
    10 970,00 €
    1 * 1 ST
       10 970,00 €
  • SAMP1230C-1500
    Triple style sampler
    Triple style sampler
    1500 mm
    8
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12 860,00 €
    12 860,00 €
    1 * 1 ST
       12 860,00 €
  • SAMP1230C-1800
    Triple style sampler
    Triple style sampler
    1800 mm
    10
    25 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14 210,00 €
    14 210,00 €
    1 * 1 ST
       14 210,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,2 ml
    Sample insert, stainless steel, 0,2 ml
    SAMP1230T-02
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,9 ml
    Sample insert, stainless steel, 0,9 ml
    SAMP1230T-09
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,0 ml
    Sample insert, stainless steel, 1,0 ml
    SAMP1230T-10
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,1 ml
    Sample insert, stainless steel, 1,1 ml
    SAMP1230T-11
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,3 ml
    Sample insert, stainless steel, 1,3 ml
    SAMP1230T-13
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,2 ml
    Sample insert, stainless steel, 1,2 ml
    SAMP1230T-12
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,4 ml
    Sample insert, stainless steel, 1,4 ml
    SAMP1230T-14
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,5 ml
    Sample insert, stainless steel, 1,5 ml
    SAMP1230T-15
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,6 ml
    Sample insert, stainless steel, 1,6 ml
    SAMP1230T-16
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,7 ml
    Sample insert, stainless steel, 1,7 ml
    SAMP1230T-17
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,8 ml
    Sample insert, stainless steel, 1,8 ml
    SAMP1230T-18
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 1,9 ml
    Sample insert, stainless steel, 1,9 ml
    SAMP1230T-19
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,0 ml
    Sample insert, stainless steel, 2,0 ml
    SAMP1230T-20
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,2 ml
    Sample insert, stainless steel, 2,2 ml
    SAMP1230T-22
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,3 ml
    Sample insert, stainless steel, 2,3 ml
    SAMP1230T-23
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,4 ml
    Sample insert, stainless steel, 2,4 ml
    SAMP1230T-24
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,5 ml
    Sample insert, stainless steel, 2,5 ml
    SAMP1230T-25
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,6 ml
    Sample insert, stainless steel, 2,6 ml
    SAMP1230T-26
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,7 ml
    Sample insert, stainless steel, 2,7 ml
    SAMP1230T-27
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,9 ml
    Sample insert, stainless steel, 2,9 ml
    SAMP1230T-29
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,0 ml
    Sample insert, stainless steel, 3,0 ml
    SAMP1230T-30
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,2 ml
    Sample insert, stainless steel, 3,2 ml
    SAMP1230T-32
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,3 ml
    Sample insert, stainless steel, 3,3 ml
    SAMP1230T-33
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,1 ml
    Sample insert, stainless steel, 3,1 ml
    SAMP1230T-31
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,4 ml
    Sample insert, stainless steel, 3,4 ml
    SAMP1230T-34
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,5 ml
    Sample insert, stainless steel, 3,5 ml
    SAMP1230T-35
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,6 ml
    Sample insert, stainless steel, 3,6 ml
    SAMP1230T-36
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,8 ml
    Sample insert, stainless steel, 3,8 ml
    SAMP1230T-38
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,9 ml
    Sample insert, stainless steel, 3,9 ml
    SAMP1230T-39
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 4,0 ml
    Sample insert, stainless steel, 4,0 ml
    SAMP1230T-40
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 3,7 ml
    Sample insert, stainless steel, 3,7 ml
    SAMP1230T-37
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,3 ml
    Sample insert, stainless steel, 0,3 ml
    SAMP1230T-03
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,4 ml
    Sample insert, stainless steel, 0,4 ml
    SAMP1230T-04
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,5 ml
    Sample insert, stainless steel, 0,5 ml
    SAMP1230T-05
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,7 ml
    Sample insert, stainless steel, 0,7 ml
    SAMP1230T-07
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,6 ml
    Sample insert, stainless steel, 0,6 ml
    SAMP1230T-06
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,8 ml
    Sample insert, stainless steel, 2,8 ml
    SAMP1230T-28
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, blank
    Sample insert, stainless steel, blank
    SAMP1230T-00
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,1 ml
    Sample insert, stainless steel, 0,1 ml
    SAMP1230T-01
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 0,8 ml
    Sample insert, stainless steel, 0,8 ml
    SAMP1230T-08
    Log in for availability
    1 * 1 ST - 152,00 €
Product Information
  • Catalog #
    Availability
    Price
  • Sample insert, stainless steel, 2,1 ml
    Sample insert, stainless steel, 2,1 ml
    SAMP1230T-21
    Log in for availability
    1 * 1 ST - 152,00 €