Racks and Boxes for Microcentrifuge Tubes
Supplier: GLW
Certificates
About this item
Styro racks are made of heavy expanded polystyrene which are used in freezer down to -90 °C. System box is made of cardboard and is designed to securely enclose two styro rack boxes.
- Not autoclavable
- For 1,5 and 2,0 ml tubes up to 12 mm diameter
Frequently Bought Together