Orders LinkContactUsLinkComponent
Inlet liners
Supplier:   PerkinElmer
Inlet liners
Supplier:  PerkinElmer
Certificates
26 Options Available Below
Product Details & Documents
Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.
  • Liners packed with wool help to trap non-volatile material
Product Family Options
Programmed temperature split/splitless (PSS) injector liners
Product Information
  • Ext.Ø
    Ø int.
    Length
    Availability
    Price
  • PERCN6101703
    Liner/hour Glass for POC injector
    Liner/hour Glass for POC injector
    4 mm
    2,4 mm
    19,05 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
  • PERCN6121006
    Quartz liner for splitless operation
    Quartz liner for splitless operation
    4 mm
    1 mm
    86,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 117,00 €
    117,00 €
    1 * 1 ST
       117,00 €
  • PERCN6121004
    Quartz liner for split operation
    Quartz liner for split operation
    4 mm
    2 mm
    86,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,20 €
    39,20 €
    1 * 1 ST
       39,20 €
  • PERCN6502000
    Siltek deactivated glass PSS liner
    Siltek deactivated glass PSS liner
    4 mm
    1 mm
    86,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 020,00 €
    1 020,00 €
    1 * 1 ST
       1 020,00 €
  • PERCN6502002
    Siltek deactivated glass PSS liner for split operation
    Siltek deactivated glass PSS liner for split operation
    4 mm
    2 mm
    86,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 822,00 €
    822,00 €
    1 * 1 ST
       822,00 €
  • PERCN9302949
    Glass injector liner
    Glass injector liner
    4 mm
    2 mm
    86,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
  • PERCN6101539
    On-column glass liner
    On-column glass liner
    4 mm
    2,4 mm
    86,2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 129,00 €
    129,00 €
    1 * 1 ST
       129,00 €
  • PERC03300905
    Silanized Glass Wool
    Silanized Glass Wool
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 104,00 €
    104,00 €
    1 * 1 ST
       104,00 €
Capillary split/splitless injector liners
Product Information
  • Ext.Ø
    Ø int.
    Length
    Availability
    Price
  • PERCN6502011
    Clarus cup split glass liner
    Clarus cup split glass liner
    6 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 050,00 €
    1 050,00 €
    1 * 1 ST
       1 050,00 €
  • PERCN6502012
    Clarus cyclosplitter glass liner
    Clarus cyclosplitter glass liner
    6 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 2 620,00 €
    2 620,00 €
    1 * 5 ST
       2 620,00 €
  • PERCN6502006
    Clarus splitless glass liner
    Clarus splitless glass liner
    6,2 mm
    1 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 489,00 €
    489,00 €
    1 * 1 ST
       489,00 €
  • PERCN6502005
    Cyclo double gooseneck liner for split operation
    Cyclo double gooseneck liner for split operation
    6,2 mm
    4 mm
    92,1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 637,00 €
    637,00 €
    1 * 1 ST
       637,00 €
  • PERCN6101372
    Glass liner for splitless operation
    Glass liner for splitless operation
    6,2 mm
    2 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,10 €
    89,10 €
    1 * 1 ST
       89,10 €
  • PERCN6101052
    Glass liner for split operation
    Glass liner for split operation
    6,2 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,20 €
    77,20 €
    1 * 1 ST
       77,20 €
  • PERCN6121002
    Quartz liner for splitless operation
    Quartz liner for splitless operation
    6,2 mm
    2 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,10 €
    51,10 €
    1 * 1 ST
       51,10 €
  • PERCN6121001
    Quartz liner for split operation
    Quartz liner for split operation
    6,2 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 77,20 €
    77,20 €
    1 * 1 ST
       77,20 €
  • PERCN6502004
    Siltek deactivated double gooseneck glass liner
    Siltek deactivated double gooseneck glass liner
    6,2 mm
    2 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 570,00 €
    2 570,00 €
    1 * 1 ST
       2 570,00 €
  • PERCN6502003
    Siltek deactivated double gooseneck glass liner
    Siltek deactivated double gooseneck glass liner
    6,2 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 420,00 €
    420,00 €
    1 * 1 ST
       420,00 €
  • PERCN6502009
    Split glass liner (with wool)
    Split glass liner (with wool)
    6 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 373,00 €
    373,00 €
    1 * 5 ST
       373,00 €
  • PERCN6121020
    Split glass liner siltek deactivated surface liner
    Split glass liner siltek deactivated surface liner
    6,2 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 630,00 €
    630,00 €
    1 * 1 ST
       630,00 €
  • PERCN6502010
    Split siltek deactivated glass liner (with wool)
    Split siltek deactivated glass liner (with wool)
    6 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 579,00 €
    579,00 €
    1 * 1 ST
       579,00 €
  • PERCN6502007
    Splitless liner
    Splitless liner
    5 mm
    2 mm
    100 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 325,00 €
    325,00 €
    1 * 1 ST
       325,00 €
Packed column injector liners
Product Information
  • Ext.Ø
    Ø int.
    Length
    Availability
    Price
  • PERCN6502017
    Cyclo uniliner
    Cyclo uniliner
    6,2 mm
    4
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 601,00 €
    601,00 €
    1 * 1 ST
       601,00 €
  • PERCN6121022
    Drilled uniliner (hole on top)
    Drilled uniliner (hole on top)
    6,2 mm
    4 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 4 110,00 €
    4 110,00 €
    1 * 1 ST
       4 110,00 €
  • PERCN6101375
    Wide-bore column glass liner
    Wide-bore column glass liner
    4 mm
    6 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,10 €
    89,10 €
    1 * 1 ST
       89,10 €
  • PERCN6121003
    Wide-bore column on/off quartz liner
    Wide-bore column on/off quartz liner
    4 mm
    6 mm
    92,1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 109,00 €
    109,00 €
    1 * 1 ST
       109,00 €