Multi-mode columns, Asahipak GS-HQ series
Supplier: Resonac
Certificates
About this item
SEC is the main separation mode. Depending on the eluent selected, the column adds multimode features of reversed phase, HILIC and ion exchange modes to SEC.
- Suitable for the separation of peptides or nucleic acids with similar molecular weights
- Suitable for desalting samples or substituting buffer in protein analysis
* Estimated value
Specifications
- Catalog No:
- 554-1976
- 554-1972
- 554-1973
- Description:
- Asahipak GS-2G 7B
- Asahipak GS-220 HQ
- Asahipak GS-320 HQ
- Max. pressure:
- 6,0 MPa (60 bar)
- 5,0 MPa (50 bar)
- pH range:
- 2 - 12
- 2 - 9
- 2 - 12
- Temperature limit:
- 4 - 60 °C
- 4 - 60 °C
- 4 - 60 °C
Specifications
Frequently Bought Together