Order Entry
Austria
ContactUsLinkComponent
General-purpose thermometer
General-purpose thermometer
  ALLA52100RC110-QP
undefined
General-purpose thermometer
  ALLA52100RC110-QP

 

  • Ø:
    8,0 - 10,0 mm
  • Immersion depth:
    Total
  • Description:
    Internal scale thermometers
  • Length:
    260 mm
  • Division:
    1,0 °C
  • Filling:
    Red
  • Pk:
    1
  • Range:
    −10...+110 °C

 

 

Until 10th October 2017 it is still allowed to use mercury thermometers if they are exclusively intended to be used for tests as per standards which require the usage of mercury thermometers.