Orders LinkContactUsLinkComponent
Ubbelohde viscometers for automatic measurements
Ubbelohde viscometers for automatic measurements
Supplier:   SI Analytics
Ubbelohde viscometers for automatic measurements
Supplier:  SI Analytics
Certificates
Ubbelohde viscometers for automatic measurements
Ubbelohde viscometers for automatic measurements
9 Options Available Below
Product Details & Documents
Viscometers with suspended ball level for determination of relative or absolute kinematic viscosity.
The additional filling and cleaning tube and glass threads ensure safe operational use.
Certifications: ISO/DIS 3105, ASTM D2515, ASTM D446
Specifications for Product Family
Specifications
  • Catalog No:
  • SCOI285402005
  • SCOI285402013
  • SCOI285402021
  • SCOI285402038
  • SCOI285402046
  • SCOI285402054
  • SCOI285402062
  • SCOI285402079
  • SCOI285402087
  • Length:
  • 290 mm
  • 290 mm
  • 290 mm
  • 290 mm
  • 290 mm
  • 290 mm
  • 290 mm
  • 290 mm
  • 290 mm
  • Volume:
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
  • 15 − 22 ml
Specifications
Product Family Options
Product Information
  • Range
    Constant
    Length
    Ø int.
    Volume
    Availability
    Price
  • 0,35 - 1 mm²/s
    0,001 K
    290 mm
    0,24 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402005
    Length
    290 mm
    Volume
    15 − 22 ml
  • 1 - 5 mm²/s
    0,005 K
    290 mm
    0,46 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402013
    Length
    290 mm
    Volume
    15 − 22 ml
  • 0,6 - 3 mm²/s
    0,003 K
    290 mm
    0,36 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402021
    Length
    290 mm
    Volume
    15 − 22 ml
  • 2 - 10 mm²/s
    0,01 K
    290 mm
    0,58 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402038
    Length
    290 mm
    Volume
    15 − 22 ml
  • 6 - 30 mm²/s
    0,03 K
    290 mm
    0,78 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402046
    Length
    290 mm
    Volume
    15 − 22 ml
  • 20 - 100 mm²/s
    0,1 K
    290 mm
    1,03 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402054
    Length
    290 mm
    Volume
    15 − 22 ml
  • 60 - 300 mm²/s
    0,3 K
    290 mm
    1,36 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402062
    Length
    290 mm
    Volume
    15 − 22 ml
  • 200 - 1000 mm²/s
    1 K
    290 mm
    1,83 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402079
    Length
    290 mm
    Volume
    15 − 22 ml
  • 600 - 3000 mm²/s
    3 K
    290 mm
    2,43 mm
    15 − 22 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 629,00 €
    629,00 €
    1 * 1 ST
       629,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    SCOI285402087
    Length
    290 mm
    Volume
    15 − 22 ml