Special pH electrodes
Supplier: CONSORT
Jump to:
About this item
3 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- TypepHTemp.MaterialElectrolyteDiaphr.Ø×LConnectionForAvailabilityPrice
- CONSSP21XSpeciality pH electrodeSpeciality pH electrodeSP21X0 - 140...+100 °CGlassRefillableSingle junction12×110 mmS7General purposesLog in for availability1 * 1 ST
- CONSSP28XSpeciality pH electrodeSpeciality pH electrodeSP28X, micro0 - 140...+80 °CGlassRefillableSingle junction4×130 mmS7Micro volumes (min. 10 mm depth), test tubesLog in for availability1 * 1 ST207,00 €
- CONSSP29XSpeciality pH electrodeSpeciality pH electrodeSingle junction, refillable0 - 140...+100 °CGlassRefillableSingle junction8×250 mmS7Tall form laboratory flasks, micro volumesLog in for availability1 * 1 ST369,00 €