Orders LinkContactUsLinkComponent
Cooling racks
Cooling racks
Supplier:   GLW
Cooling racks
Supplier:  GLW
Certificates
Cooling racks
4 Options Available Below
Product Details & Documents
These cooling racks are made of polypropylene for test tubes that require continued cooling.
  • Sturdy construction
  • Autoclavable up to +121 °C
  • Cover must be ordered seperately
Remove the insert, add chrushed ice, reinsert and start working. Use in freezer down to −90° C
Product Family Options
Product Information
  • No. of Holes
    W×D×H
    Availability
    Price
  • GLWAG01
    Cooling rack, natural, PP, for tubes up to 0,5 ml
    Cooling rack, natural, PP, for tubes up to 0,5 ml
    100
    130×130×57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,56 €
    9,56 €
    1 * 1 ST
       9,56 €
  • GLWAG01S
    Cooling rack, PP, black, for tubes up to 0,5 ml
    Cooling rack, PP, black, for tubes up to 0,5 ml
    100
    130×130×57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,68 €
    9,68 €
    1 * 1 ST
       9,68 €
  • GLWAG02
    Cooling rack, PP, natural, for tubes up to 1,5 or 2,0 ml
    Cooling rack, PP, natural, for tubes up to 1,5 or 2,0 ml
    64
    130×130×57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,50 €
    9,50 €
    1 * 1 ST
       9,50 €
  • GLWAG02S
    Cooling rack, PP, black, for tubes up to 1,5 or 2,0 ml
    Cooling rack, PP, black, for tubes up to 1,5 or 2,0 ml
    64
    130×130×57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,62 €
    9,62 €
    1 * 1 ST
       9,62 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cover for cooling racks, PP, natural, autoclavable
    Cover for cooling racks, PP, natural, autoclavable
    GLWAGD1
    Log in for availability
    1 * 1 ST - 1,96 €
Product Information
  • Catalog #
    Availability
    Price
  • Cover for cooling racks, PP, black, autoclavable
    Cover for cooling racks, PP, black, autoclavable
    GLWAGD1S
    Log in for availability
    1 * 1 ST - 2,02 €