ContactUsLinkComponent
Reversed phase chromatography columns, silica-based C18M
Reversed phase chromatography columns, silica-based C18M
Supplier:  Resonac
Certificates
undefined
Reversed phase chromatography columns, silica-based C18M
Supplier:  Resonac
Specifications
  • Catalog No:
  • SHOWF6650041
  • SHOWF6650040
  • Description:
  • Silica C18M 4E
  • Silica C18M 4D
  • Carbon load:
  • 16%
  • 16%
  • Endcapped:
  • Yes
  • Yes
  • Max. pressure:
  • 20 MPa (200 bar)
  • 20 MPa (200 bar)
  • pH range:
  • 2 - 7,5
  • 2 - 7,5
  • Temperature limit:
  • 20 - 60 °C
  • 20 - 60 °C
  • USP code:
  • L1
  • L1