Orders LinkContactUsLinkComponent
Precision thermometers, Safe BIO-TEMP®
Supplier:   Alla France
Precision thermometers, Safe BIO-TEMP®
Supplier:  Alla France
Certificates
Precision thermometers, Safe BIO-TEMP®
Precision thermometers, Safe BIO-TEMP®
3 Options Available Below
Product Details & Documents
Mercury-free thermometers with yellow back glass.
  • PTFE coated thermometer filled with environmentally safe blue organic filling liquid, for extra security
  • A high density PE base houses the reservoir of the thermometer, acting as a type of thermal brake
  • Instrument can be free-standing or attached
 : Supplied with adhesive strip and magnet, in a recyclable sturdy plastic concertina box, ideal for secure storage when not in use, and with a conformity certificate, (on request) can be supplied with 'EU' Traceable calibration certificate.
Product Family Options
Product Information
  • Range
    Accuracy
    Division
    Length
    Filling
    Immersion depth
    Ø
    Availability
    Price
  • −80...+20 °C
    ±1,0 °C
    1,0 °C
    150 mm
    Blue organic liquid
    Total
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
  • −5...+30 °C
    ±0,5 °C
    0,5 °C
    150 mm
    Blue organic liquid
    Total
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €
  • +20…+50 °C
    ±0,5 °C
    0,5 °C
    150 mm
    Blue organic liquid
    Total
    6 - 7 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 162,00 €
    162,00 €
    1 * 1 ST
       162,00 €