Orders LinkContactUsLinkComponent
OpenLAB CDS EZChrom edition
OpenLAB CDS EZChrom edition
Supplier:   VWR Collection
OpenLAB CDS EZChrom edition
Supplier:  VWR Collection
Certificates
OpenLAB CDS EZChrom edition
OpenLAB CDS EZChrom edition
6 Options Available Below
Product Details & Documents
Scalable, compliant chromatography software platform with multi-instrument control. OpenLAB CDS EZChrom Edition is part of the Agilent software suite OpenLAB for analytical laboratories and enterprise. Its multi-instrument control capabilities, scalability and compliance are ideal to fulfil the requirements of today's chromatography laboratories, and to optimise their efficiency. Central administration functions have been arranged in the OpenLAB Shared Services/Control Panel module while keeping the existing user-interface and workflows for the EZChrom Elite™ chromatography data system. Many exciting functions are now available, from the Result Package mode and Result Review/Result Sequence mode to the new Intelligent Reporting mode. OpenLAB CDS EZChrom Edition can be easily complemented by further software modules belonging to the OpenLAB software suite.
  • Scalable from workstations to fully featured Client/Server systems
  • Configurable compliance
  • Multi-instrument control (many different LC and GC instrument control drivers available)
  • Intelligent reporting
  • Result Package mode
For further information and additional articles, please contact our specialists at Avantor or email [email protected].
Product Family Options
Product Information
  • Availability
    Price
  • 906-0145
    OpenLAB CDS EZChrom edition including 1 year software maintenance agreement + Hitachi LC control
    OpenLAB CDS EZChrom edition including 1 year software maintenance agreement + Hitachi LC control
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 9 690,00 €
    9 690,00 €
    1 * 1 SET
       9 690,00 €
  • 906-0146
    OpenLAB CDS EZChrom edition including 1 year software maintenance agreement + Hitachi LC control + Hitachi DAD
    OpenLAB CDS EZChrom edition including 1 year software maintenance agreement + Hitachi LC control + Hitachi DAD
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 12 760,00 €
    12 760,00 €
    1 * 1 SET
       12 760,00 €
  • 906-0147
    OpenLab CDS EZChrom edition licence for additional Hitachi HPLC on the same PC
    OpenLab CDS EZChrom edition licence for additional Hitachi HPLC on the same PC
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 4 390,00 €
    4 390,00 €
    1 * 1 SET
       4 390,00 €
  • 906-0148
    OpenLab CDS EZChrom edition licence for additional Hitachi HPLC with diode array on the same PC
    OpenLab CDS EZChrom edition licence for additional Hitachi HPLC with diode array on the same PC
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 7 460,00 €
    7 460,00 €
    1 * 1 SET
       7 460,00 €
  • 906-0097
    OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software renewal of 1 year software subscription
    OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software renewal of 1 year software subscription
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 771,00 €
    771,00 €
    1 * 1 ST
       771,00 €
  • 906-0093
    OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software including 1 year software subscription
    OpenLAB CDS EZChrom edition workstation, basic workstation chromatography software including 1 year software subscription
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 5 300,00 €
    5 300,00 €
    1 * 1 ST
       5 300,00 €