Drum pumps, mains operated
Supplier: BURKLE
Certificates
About this item
These are light- and high-performance pump to transfer small quantities of watery media up to 300 mPas from canisters, barrels and narrow necked containers from 32 mm opening diameter. Laboratory pumps, with splash water protected motor 230 V/200 W (IP 44), with 1,5 m PVC-hose, PP nozzle, 2 hose clips, discharge spout, wall bracket.
- Made up of PP
- Built-in safety key as on/off switch
- Seal-less pump mechanism for acids and alkalis
- Thread connection 1" with hose connector 3/4"
Frequently Bought Together