Retort bases, with M10 thread
Pressed steel finished in matt black textured acrylic paint, threaded M10 × 1,5 mm.
: Retort rods are not included, they can be ordered separately under "Complementary products".
Frequently Bought Together
Pressed steel finished in matt black textured acrylic paint, threaded M10 × 1,5 mm.