Orders LinkContactUsLinkComponent
Safety cans | Avantor
Safety cans
Safety cans
Certificates
Safety cans
14 Options Available Below
Product Details & Documents
High quality, chemically resistant stainless steel with PTFE gaskets.
  • Easy handling, ergonomic design and high stability
  • Flame traps and explosion-proof
  • Pressure relief valve (vent at about 0,3 bar)
  • Self-closing metering devices
  • Moisteners for safe cleaning with solvents
For safe and comfortable handling of small amounts of flammable liquids.
Dokumente
Product Family Options
Product Information
  • Capacity
    Availability
    Price
  • 119-0103
    Safety can with screw cap 1¼"
    Safety can with screw cap 1¼"
    1 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 156,00 €
    156,00 €
    1 * 1 ST
       156,00 €
  • 119-2000
    Safety can with screw cap 1¼", electro-polished
    Safety can with screw cap 1¼", electro-polished
    1 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 165,00 €
    165,00 €
    1 * 1 ST
       165,00 €
  • 119-0106
    Safety can with screw cap 1¼"
    Safety can with screw cap 1¼"
    2 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 172,00 €
    172,00 €
    1 * 1 ST
       172,00 €
  • 119-2042
    Safety can with screw cap 1¼", electro-polished
    Safety can with screw cap 1¼", electro-polished
    2 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 181,00 €
    181,00 €
    1 * 1 ST
       181,00 €
  • 119-0114
    Safety can with screw cap 1¼“
    Safety can with screw cap 1¼“
    5 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 201,00 €
    201,00 €
    1 * 1 ST
       201,00 €
  • 119-2005
    Safety can with screw cap 1¼“, electro-polished
    Safety can with screw cap 1¼“, electro-polished
    5 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 212,00 €
    212,00 €
    1 * 1 ST
       212,00 €
  • 119-0102
    Safety can with metering device 1¼"
    Safety can with metering device 1¼"
    1 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 191,00 €
    191,00 €
    1 * 1 ST
       191,00 €
  • 119-0101
    Safety can with metering device 1¼", electro-polished
    Safety can with metering device 1¼", electro-polished
    1 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 202,00 €
    202,00 €
    1 * 1 ST
       202,00 €
  • 119-0105
    Safety can with metering device 1¼"
    Safety can with metering device 1¼"
    2 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 207,00 €
    207,00 €
    1 * 1 ST
       207,00 €
  • 119-0104
    Safety can with metering device 1¼", electro-polished
    Safety can with metering device 1¼", electro-polished
    2 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 219,00 €
    219,00 €
    1 * 1 ST
       219,00 €
  • 119-0107
    Safety can with metering device 1¼"
    Safety can with metering device 1¼"
    5 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
  • 119-0171
    Safety can with metering device 1¼“, electro-polished
    Safety can with metering device 1¼“, electro-polished
    5 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 245,00 €
    245,00 €
    1 * 1 ST
       245,00 €
  • 119-0112
    Moistener
    Moistener
    1 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 225,00 €
    225,00 €
    1 * 1 ST
       225,00 €
  • 119-0113
    Moistener
    Moistener
    2 l
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 240,00 €
    240,00 €
    1 * 1 ST
       240,00 €
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Flame trap
    Flame trap
    119-0170
    All Rötzmeier safety cans, moisteners and safety transportation cans
    Log in for availability
    1 * 1 ST - 17,20 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Soaking tray with pump for safety can, 1 l
    Soaking tray with pump for safety can, 1 l
    119-0139
    119-2000, -0103, -0101, -0102
    Log in for availability
    1 * 1 ST - 132,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Soaking tray with pump for safety can, 2 l
    Soaking tray with pump for safety can, 2 l
    119-0140
    119-2042, -0106, -0104, -0105
    Log in for availability
    1 * 1 ST - 134,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Screw cap 1¼" with pressure control valve
    Screw cap 1¼" with pressure control valve
    119-0144
    All Rötzmeier safety cans and moisteners
    Log in for availability
    1 * 1 ST - 53,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Self-closing metering device 1¼" with ventilation
    Self-closing metering device 1¼" with ventilation
    119-2390
    All Rötzmeier safety cans and moisteners
    Log in for availability
    1 * 1 ST - 77,00 €
Empfehlungen
Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.
Andernfalls erhalten Sie allgemeine Empfehlungen.