Orders LinkContactUsLinkComponent
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
Supplier:   COWIE
Stopcocks, straight through, with tip
Supplier:  COWIE
Certificates
Stopcocks, straight through, with tip
Stopcocks, straight through, with tip
4 Options Available Below
Product Details & Documents
Made of PTFE.
  • Available with PP or glass tip
  • Very good chemical resistance
  • Very good temperature resistance
Product Family Options
Tips for burettes
Product Information
  • Hole Ø
    Availability
    Price
  • PP
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,10 €
    16,10 €
    1 * 10 ST
       16,10 €
  • Glass
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 16,10 €
    16,10 €
    1 * 10 ST
       16,10 €
Product Information
  • Hole Ø
    Availability
    Price
  • 661-1181
    with polypropylene tip
    with polypropylene tip
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,40 €
    24,40 €
    1 * 1 ST
       24,40 €
  • COWI016.9203-B
    with glass tip
    with glass tip
    2 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,40 €
    24,40 €
    1 * 1 ST
       24,40 €