Tubing connectors, SVS, bent
Catalog # WITG0.185.001
Supplier: witeg Labortechnik
Specifications
- Thread:GL 14
- Tubing int.Ø:8 mm
- Material:PP
- Pk:1
Specifications
About this item
Tubing connection consisting of screw cap, hose connection with silicone seal and threaded tubes.
- Very good chemical resistance
- Very good temperature resistance