Order Entry
Austria
ContactUsLinkComponent
Screw caps for thread connection
Screw caps for thread connection
Catalog # WITG0.176.032
Supplier:  witeg Labortechnik
undefined
Screw caps for thread connection
Catalog # WITG0.176.032
Supplier:  witeg Labortechnik

Specifications

  • Hole Ø:
    20 mm
  • Thread:
    GL 32
  • Pk:
    1

Specifications

About this item

PBT, red without seals.

  • For DIN GL threads
  • Heat resistant up to 180 °C
  • Autoclavable, sterilisable with hot air

With central hole.