ContactUsLinkComponent
Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Certificates
undefined
Vortex mixers, large capacity, Multi Vortex-Genie® and Vortex Genie® Max
Specifications
  • Catalog No:
  • 444-1299
  • 444-1292
  • Model:
  • Vortex-Genie Max
  • Multi Vortex-Genie
  • Shaking motion:
  • Orbital
  • Orbital
  • Orbit:
  • 3 mm
  • 4 mm
  • Speed range:
  • 500 - 1950 min⁻¹
  • 200 - 950 min⁻¹
  • Max. Ø of tubes:
  • 20 mm
  • 29 mm
  • W×D×H:
  • 270×310×195 mm
  • 270×310×195 mm
  • Weight:
  • 10,4 kg
  • 11,3 kg