Pump tubing, Tygon®
Thermoplastic soft PVC, transparent.
- Non-toxic, non-oxidizing
- Good resistance to acids, bases and inorganic media
- Very low gas permeability, good performance life
For standard applications.
: FDA and GLP conform.
Frequently Bought Together